missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris(trimethylsilyloxy)ethylene, TCI America™

Manufacturer:  TCI America T163925G

 View more versions of this product

Catalog No. T163925G



Specifications

Specifications

Tris(trimethylsilyloxy)ethylene
25g
C11H28O3Si3
1993
FCZGHPGTZRTDNN-UHFFFAOYSA-N
1,2-bis(trimethylsilyloxy)ethenoxy-trimethylsilane
553067
Clear Liquid at 20°C
Colorless
69097-20-7
MFCD00011641
tris trimethylsilyloxy ethylene, tris trimethylsiloxy ethylene, vinyl-2-ylidenetris oxy tris trimethylsilane, 2,2,7,7-tetramethyl-4-trimethylsilyl oxy-3,6-dioxa-2,7-disilaoct-4-ene, 2,2,7,7-tetramethyl-4-trimethylsilyl oxy-3,6-dioxa-2,7-disilaoct-4-ene, 3,6-dioxa-2,7-disilaoct-4-ene, 2,2,7,7-tetramethyl-4-trimethylsilyl oxy, acmc-1b3cp, tris trimethylsiloxy ethene, tris trimethylsiloxy-ethylene, tris-trimethylsiloxy ethylene
C[Si](C)(C)OC=C(O[Si](C)(C)C)O[Si](C)(C)C
292.597
292.60
≥95.0% (GC)

Chemical Identifiers

69097-20-7
292.597
FCZGHPGTZRTDNN-UHFFFAOYSA-N
553067
C[Si](C)(C)OC=C(O[Si](C)(C)C)O[Si](C)(C)C
C11H28O3Si3
MFCD00011641
tris trimethylsilyloxy ethylene, tris trimethylsiloxy ethylene, vinyl-2-ylidenetris oxy tris trimethylsilane, 2,2,7,7-tetramethyl-4-trimethylsilyl oxy-3,6-dioxa-2,7-disilaoct-4-ene, 2,2,7,7-tetramethyl-4-trimethylsilyl oxy-3,6-dioxa-2,7-disilaoct-4-ene, 3,6-dioxa-2,7-disilaoct-4-ene, 2,2,7,7-tetramethyl-4-trimethylsilyl oxy, acmc-1b3cp, tris trimethylsiloxy ethene, tris trimethylsiloxy-ethylene, tris-trimethylsiloxy ethylene
1,2-bis(trimethylsilyloxy)ethenoxy-trimethylsilane
Safety and Handling

Safety and Handling

TSCA : No

Documents