Please login to your online account to display your discounted pricing

Tris(trimethylsilyloxy)ethylene, 95%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  216010050

 View more versions of this product

Catalog No. AC216010050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 292.59
Physical Form Liquid
Quantity 5g
Color Clear colorless to yellow
Fieser 08,523; 09,512
Density 0.8850g/mL
Packaging Glass bottle
Linear Formula [(CH3)3SiO]2C=CHOSi(CH3)3
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C11H28O3Si3
Chemical Name or Material Tris(trimethylsilyloxy)ethylene
MDL Number MFCD00011641
CAS 69097-20-7
Assay Percent Range 95%
Refractive Index 1.4190 to 1.4220 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Warning

Specifications

Chemical Name or Material Tris(trimethylsilyloxy)ethylene
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C11H28O3Si3
Linear Formula [(CH3)3SiO]2C=CHOSi(CH3)3
Formula Weight 292.59
MDL Number MFCD00011641
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

SDS & Safety Documents