Please login to your online account to display your discounted pricing

Tris(trimethylsilyl) phosphite, 92%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC428520050

 View more versions of this product

Catalog No. AC428520050

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥90%
Formula Weight 298.54
Quantity 5g
Color Clear colorless
Density 0.8930g/mL
Chemical Name or Material Tris(trimethylsilyl) phosphite
Molecular Formula C9H27O3PSi3
Linear Formula P[OSi(CH3)3]3
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Assay Percent Range 92%
MDL Number MFCD00015588
CAS 1795-31-9
Refractive Index 1.4080 to 1.4110 (20°C, 589nm)
Health Hazard 1 Warning

Specifications

Chemical Name or Material Tris(trimethylsilyl) phosphite
Assay ≥90%
Assay Percent Range 92%
Molecular Formula C9H27O3PSi3
Linear Formula P[OSi(CH3)3]3
Formula Weight 298.54
MDL Number MFCD00015588
Accessories

Warning

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents