Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(trimethylsilyl)phosphine

Chemical Identifiers

Chemical Name or Material Tris(trimethylsilyl)phosphine
CAS 15573-38-3
Formula Weight 250.55
MDL Number MFCD00015487
UN Number UN2845
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3021803

  SDS
alfa aesar™
3021803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA30218MD

  SDS
alfa aesar™
30218MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 250.55
Chemical Name or Material Tris(trimethylsilyl)phosphine
Physical Form Liquid
MDL Number MFCD00015487
CAS 15573-38-3
UN Number UN2845
Refractive Index 1.5020
Health Hazard 1 Danger
Density 0.863g/mL
Health Hazard 2 GHS H Statement
H250-H252-H315-H319
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes skin irritation.
Causes serious eye irritation.
Melting Point 24°C
Boiling Point 102° to 105°C (16mmHg)
Odor Pungent
Quantity 1g
Health Hazard 3 GHS P Statement
P210-P222-P280-P305+P351+P338-P362-P422a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not allow contact with air.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store contents under inert gas.
Sensitivity Air & Moisture Sensitive
DOT Information Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P222-P280-P305+P351+P338-P362-P422a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not allow contact with air.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store contents under inert gas.

GHS H Statement
H250-H252-H315-H319
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes skin irritation.
Causes serious eye irritation.