missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(trimethylsilyl)phosphine

Manufacturer:  Alfa Aesar™ 3021803

 View more versions of this product

Catalog No. AA3021803



Specifications

Specifications

Tris(trimethylsilyl)phosphine
Pungent
Air & Moisture Sensitive
24°C
15573-38-3
MFCD00015487
tris trimethylsilyl phosphine, tris trimethylsilyl phosphane, phosphine, tris trimethylsilyl, tris trimethylsilyl phosphine, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl disilaphosphane, tris trimethylsilyl phosphine in hexane, pubchem6456, acmc-1btki, tris-trimethylsilyl phosphine, phosphine,tris trimethylsilyl
C[Si](C)(C)P([Si](C)(C)C)[Si](C)(C)C
250.544
250.55
0.863g/mL
1.5020
102°C to 105°C (16mmHg)
1g
C9H27PSi3
UN2845
OUMZKMRZMVDEOF-UHFFFAOYSA-N
tris(trimethylsilyl)phosphane
272683
Liquid

Chemical Identifiers

15573-38-3
250.544
OUMZKMRZMVDEOF-UHFFFAOYSA-N
272683
C[Si](C)(C)P([Si](C)(C)C)[Si](C)(C)C
C9H27PSi3
MFCD00015487
tris trimethylsilyl phosphine, tris trimethylsilyl phosphane, phosphine, tris trimethylsilyl, tris trimethylsilyl phosphine, 1,1,1,3,3,3-hexamethyl-2-trimethylsilyl disilaphosphane, tris trimethylsilyl phosphine in hexane, pubchem6456, acmc-1btki, tris-trimethylsilyl phosphine, phosphine,tris trimethylsilyl
tris(trimethylsilyl)phosphane
Safety and Handling

Safety and Handling

GHS H Statement
H250-H252-H315-H319
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P210-P222-P280-P305+P351+P338-P362-P422a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not allow contact with air.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Store contents under inert gas.

Danger

DOTInformation : Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.

SDS Documents