missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tris(trimethylsilyl) Borate 97.0+%, TCI America™
Supplier: TCI America T254325G
Specifications
Tris(trimethylsilyl) Borate | |
-35°C | |
180°C | |
MFCD00051588 | |
1993 | |
YZYKZHPNRDIPFA-UHFFFAOYSA-N | |
tris(trimethylsilyl) borate | |
78020 | |
≥97.0% (T) |
4325-85-3 | |
Yellow | |
C9H27BO3Si3 | |
25 g | |
tris trimethylsilyl borate, tris trimethylsiloxy boron, silanol, trimethyl-, triester with boric acid h3bo3, tritrimethylsilyl borate, borane, tris trimethylsiloxy, silanol, 1,1,1-trimethyl-, 1,1',1-triester with boric acid h3bo3, acmc-209jtk, tris trimethylsiloxy borane, boric acid, 3tms derivative | |
B(O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C | |
278.377 | |
278.38 | |
Liquid |
Chemical Identifiers
4325-85-3 | |
278.377 | |
YZYKZHPNRDIPFA-UHFFFAOYSA-N | |
78020 | |
B(O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C |
C9H27BO3Si3 | |
MFCD00051588 | |
tris trimethylsilyl borate, tris trimethylsiloxy boron, silanol, trimethyl-, triester with boric acid h3bo3, tritrimethylsilyl borate, borane, tris trimethylsiloxy, silanol, 1,1,1-trimethyl-, 1,1',1-triester with boric acid h3bo3, acmc-209jtk, tris trimethylsiloxy borane, boric acid, 3tms derivative | |
tris(trimethylsilyl) borate |
Safety and Handling
TSCA : Yes