missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris(trimethylsilyl) Borate, TCI America™

Manufacturer:  TCI America T2543100G

 View more versions of this product

Catalog No. T2543100G



Specifications

Specifications

Tris(trimethylsilyl) Borate
Yellow
100g
C9H27BO3Si3
1993
YZYKZHPNRDIPFA-UHFFFAOYSA-N
tris(trimethylsilyl) borate
78020
Clear Liquid at 20°C
180°C
-35°C
4325-85-3
MFCD00051588
tris trimethylsilyl borate, tris trimethylsiloxy boron, tris trimethylsilyl borate, silanol, trimethyl-, triester with boric acid h3bo3, tritrimethylsilyl borate, borane, tris trimethylsiloxy, silanol, 1,1,1-trimethyl-, 1,1',1-triester with boric acid h3bo3, acmc-209jtk, tris trimethylsiloxy borane, boric acid, 3tms derivative
B(O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C
278.377
278.38
≥97.0% (T)

Chemical Identifiers

4325-85-3
278.377
YZYKZHPNRDIPFA-UHFFFAOYSA-N
78020
B(O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C
C9H27BO3Si3
MFCD00051588
tris trimethylsilyl borate, tris trimethylsiloxy boron, tris trimethylsilyl borate, silanol, trimethyl-, triester with boric acid h3bo3, tritrimethylsilyl borate, borane, tris trimethylsiloxy, silanol, 1,1,1-trimethyl-, 1,1',1-triester with boric acid h3bo3, acmc-209jtk, tris trimethylsiloxy borane, boric acid, 3tms derivative
tris(trimethylsilyl) borate
Safety and Handling

Safety and Handling

TSCA : Yes

Documents