Please login to your online account to display your discounted pricing

Tris(trimethylsilyl) borate, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC381240250

 View more versions of this product

Catalog No. AC381240250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.0%
Physical Form Liquid
Formula Weight 278.38
Quantity 25g
Color Clear colorless
Density 0.834g/mL
Refractive Index 1.3850 to 1.3870 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
Molecular Formula C9H27BO3Si3
CAS 4325-85-3
Linear Formula B(OSi(CH3)3)
Health Hazard 1 GHS Signal Word: Warning
Chemical Name or Material Tris(trimethylsilyl) borate
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00051588
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

Specifications

Chemical Name or Material Tris(trimethylsilyl) borate
Assay ≥97.0%
Molecular Formula C9H27BO3Si3
Linear Formula B(OSi(CH3)3)
Formula Weight 278.38
MDL Number MFCD00051588
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

SDS & Safety Documents