Please login to your online account to display your discounted pricing

Tris(trimethylsilyl)amine, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Tris(trimethylsilyl)amine
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H27NSi3
Linear Formula N(Si(CH3)3)3
Formula Weight 233.58
MDL Number MFCD00047990
Synonym Nonamethyltrisilazane
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC378770050

  SDS
thermo fisher scientific
AC378770050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC378770250

  SDS
thermo fisher scientific
AC378770250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline
Formula Weight 233.58
Quantity 5g
Color Colorless
Chemical Name or Material Tris(trimethylsilyl)amine
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C9H27NSi3
Linear Formula N(Si(CH3)3)3
Packaging Glass bottle
MDL Number MFCD00047990
CAS 1586-73-8
Synonym Nonamethyltrisilazane
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Causes serious eye damage.
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Causes serious eye damage.