missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(trifluoromethylhydroxymethylene-d-camphorato)europium(III)

Manufacturer:  Alfa Aesar™ 8848103

 View more versions of this product

Catalog No. AA8848103



Specifications

Specifications

Tris (trifluoromethylhydroxymethylene-d-camphorato)europium (III)
195°C to 198°C
34830-11-0
MFCD00074803
YXJPWWGLCOPUGI-UFRADDTFSA-N
europium;(2E)-4,7,7-trimethyl-2-(2,2,2-trifluoro-1-hydroxyethylidene)bicyclo[2.2.1]heptan-3-one
71310195
Crystalline
Moisture Sensitive
1g
C36H45EuF9O6
eu tfc 3, eu facam 3, europium tris 3-trifluoromethylhydroxymethylene---camphorate, europium tris 3-trifluoromethylhydroxymethylene-+-camphorate, tris 3-trifluoromethylhydroxymethylene-d-camphorato europium iii, tris 3-trifluoroacetyl-d-camphorato europium iii, europium iii tris 3-trifluoromethylhydroxymethylene-d-camphorate
CC1(C2CCC1(C(=O)C2=C(C(F)(F)F)O)C)C.CC1(C2CCC1(C(=O)C2=C(C(F)(F)F)O)C)C.CC1(C2CCC1(C(=O)C2=C(C(F)(F)F)O)C)C.[Eu]
896.7
893.68

Chemical Identifiers

34830-11-0
896.7
YXJPWWGLCOPUGI-UFRADDTFSA-N
71310195
CC1(C2CCC1(C(=O)C2=C(C(F)(F)F)O)C)C.CC1(C2CCC1(C(=O)C2=C(C(F)(F)F)O)C)C.CC1(C2CCC1(C(=O)C2=C(C(F)(F)F)O)C)C.[Eu]
C36H45EuF9O6
MFCD00074803
eu tfc 3, eu facam 3, europium tris 3-trifluoromethylhydroxymethylene---camphorate, europium tris 3-trifluoromethylhydroxymethylene-+-camphorate, tris 3-trifluoromethylhydroxymethylene-d-camphorato europium iii, tris 3-trifluoroacetyl-d-camphorato europium iii, europium iii tris 3-trifluoromethylhydroxymethylene-d-camphorate
europium;(2E)-4,7,7-trimethyl-2-(2,2,2-trifluoro-1-hydroxyethylidene)bicyclo[2.2.1]heptan-3-one
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents