Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris-Tricine Large Precast Gel, 10-20%, 16x20cm, 18.3x20cm (back), 1mm thick, 15 well, for Bio-Rad™ systems


Manufacturer: alfa aesar™  J67573KZ

 View more versions of this product

Catalog No. AAJ67573KZ

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Boiling Point 100°C
Storage Requirements 2° to 8°C
Quantity 6 Each
Product Type Tris-Tricine Large Precast Gel
For Use With (Application) For separating small proteins from 2.3kD to 100kD in size
Odor Odorless
Format Solid
Color Colorless