Alfa Aesar™ Tris-Tricine Large Precast Gel, 10-20%, 16x20cm, 18.3x20cm (back), 1.5mm thick, 15 well, for Bio-Rad™ systems
Manufacturer:  Alfa Aesar™ J67750KZ
 View more versions of this product
Catalog No. AAJ67750KZ
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications
Specifications
100°C
For separating small proteins from 2.3kD to 100kD in size
6 Each
Odorless
Colorless
Solid
2° to 8°C
Tris-Tricine Large Precast Gel