Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(tetramethylcyclopentadienyl)neodymium(III), 99% (99.9%-Nd) (REO)

Chemical Identifiers

Chemical Name or Material Tris(tetramethylcyclopentadienyl)neodymium(III)
CAS 164528-22-7
Name Note (REO)
Molecular Formula C27H39Nd
Formula Weight 507.85
MDL Number MFCD01862468
UN Number UN1325
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2718503

  SDS
alfa aesar™
H2718503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 507.85
Chemical Name or Material Tris(tetramethylcyclopentadienyl)neodymium(III)
Name Note (REO)
Assay 99%
Assay Percent Notes (99.9%-Nd)
Molecular Formula C27H39Nd
MDL Number MFCD01862468
CAS 164528-22-7
UN Number UN1325
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H228
Flammable solid.
Flash Point 48°C (118°F)
Quantity 1g
Health Hazard 3 GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.
DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

GHS H Statement
H228
Flammable solid.