Please login to your online account to display your discounted pricing

Tris Solution, 1.0M, pH 9.0, OmniPur™, Calbiochem™, EMD Millipore

Useful in the pH range of 9.0

$75.99 - $75.99

Chemical Identifiers

CAS 7732-18-5
Chemical Name or Material Tris Solution
Concentration or Composition (by Analyte or Components) 1M Solution
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

9295100ML

 
emd millipore corp
9295100ML
Each for $75.99
Description & Specifications

Specifications

Quantity 100mL
Chemical Name or Material Tris Solution
CAS 7732-18-5
DNase None Detected
Concentration or Composition (by Analyte or Components) 1M Solution
Solubility Information Soluble in water
CAS 77-86-1
Protease None Detected
Recommended Storage Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.
CAS 7647-01-0
RNase None Detected
Safety and Handling

Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.