missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris Solution, 1.0M, pH 7.5, OmniPur™, Calbiochem™, EMD Millipore

Useful in the pH range of 7.5

$75.44 - $75.44

Chemical Identifiers

CAS 7732-18-5
Concentration or Composition (by Analyte or Components) 1M Solution
Chemical Name or Material Tris Solution
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
9285100ML Emd Millipore Corp
9285100ML
100mL Each for $75.44
Specifications Description & Specifications

Chemical Identifiers

7732-18-5
1M Solution
Tris Solution

Specifications

7732-18-5,77-86-1,7647-01-0
1M Solution
None Detected
100mL
None Detected
Soluble in water
None Detected
Tris Solution

Safety and Handling

Recommended Storage : Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.