Please login to your online account to display your discounted pricing

Tris SDS PAGE Running Buffer, FASTRun, 10X


Compatible with Tris-Glycine SDS (SDS-PAGE) chemistry whether you buy premade or pour your own (PYO) polyacrylamide gels

Manufacturer: fisher scientific company  BP881500

 View more versions of this product

Catalog No. BP881500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

CAS 77-86-1
CAS 151-21-3
CAS 7732-18-5
Purity Grade Protein Electrophoresis
Clarity clear, no visible precipitates
Physical Form Liquid
Conductivity 2 to 3 mS/cm ( 1X Solution at 25°C)
Synonym Denaturing Buffer
Packaging Poly
pH 7.7 to 8.3 (1X/10X soln. at 25°C)
Color Colorless to Light Yellow
Quantity 500mL

Specifications

CAS 77-86-1
CAS 151-21-3
CAS 7732-18-5
Purity Grade Protein Electrophoresis
Synonym Denaturing Buffer

  • Cut the time you spend waiting for your SDS PAGE gels to finish by over 70% while providing comparable or better clarity over traditional buffers
  • Novel buffer chemistry gives you better resolution, thus requiring fewer TG gel compositions to fully resolve a protein.