missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris(phenylthio)methane, TCI America™

Manufacturer:  TCI America T312625G

 View more versions of this product

Catalog No. T312625G



Specifications

Specifications

Tris(phenylthio)methane
41°C
4832-52-4
MFCD00010396
C1=CC=C(C=C1)SC(SC2=CC=CC=C2)SC3=CC=CC=C3
340.517
340.52
≥98.0% (GC)
White
25g
C19H16S3
YOQHDHPBEXTHCP-UHFFFAOYSA-N
bis(phenylsulfanyl)methylsulfanylbenzene
615706
Crystal-Powder at 20°C

Chemical Identifiers

4832-52-4
340.517
YOQHDHPBEXTHCP-UHFFFAOYSA-N
bis(phenylsulfanyl)methylsulfanylbenzene
C19H16S3
MFCD00010396
615706
C1=CC=C(C=C1)SC(SC2=CC=CC=C2)SC3=CC=CC=C3
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents