Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(pentafluorophenyl)phosphine, 97%

$67.05 - $226.01

Chemical Identifiers

Chemical Name or Material Tris(pentafluorophenyl)phosphine
CAS 1259-35-4
Molecular Formula C18F15P
Formula Weight 532.15
MDL Number MFCD00079654
Beilstein 773243
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0274803

  SDS
alfa aesar™
L0274803
1g Each for $67.05

AAL0274806

  SDS
alfa aesar™
L0274806
5g Each for $226.01
Description & Specifications

Specifications

Formula Weight 532.15
Health Hazard 1 Warning
Assay 97%
Chemical Name or Material Tris(pentafluorophenyl)phosphine
MDL Number MFCD00079654
Melting Point 115° to 119°C
Odor Odorless
Beilstein 773243
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 1259-35-4
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Quantity 1g
Molecular Formula C18F15P

Applications
Tris(pentafluorophenyl)phosphine is an additive for high voltage batteries. It acts as building blocks, coupling reagents, derivatizing reagents.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances. Handle under inert gas. Incompatible with Strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.