missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris(pentafluorophenyl)borane, TCI America™

Manufacturer:  TCI America T23131G

 View more versions of this product

Catalog No. T23131G



Specifications

Specifications

Tris(pentafluorophenyl)borane
128°C
1109-15-5
MFCD00269813
OBAJXDYVZBHCGT-UHFFFAOYSA-N
tris(2,3,4,5,6-pentafluorophenyl)borane
582056
Crystal-Powder at 20°C
Undesignated
1g
C18BF15
tris pentafluorophenyl borane, tris perfluorophenyl borane, tris pentafluorophenyl boron, tris 2,3,4,5,6-pentafluorophenyl borane, unii-i3wu5e2578, tris pentafluorophenyl-borane, borane, tris pentafluorophenyl, borane, tris 2,3,4,5,6-pentafluorophenyl, perfluorotriphenylboron, zlchem 1272
B(C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)C3=C(C(=C(C(=C3F)F)F)F)F
511.984
511.98
≥98.0% (NMR)

Chemical Identifiers

1109-15-5
511.984
OBAJXDYVZBHCGT-UHFFFAOYSA-N
582056
B(C1=C(C(=C(C(=C1F)F)F)F)F)(C2=C(C(=C(C(=C2F)F)F)F)F)C3=C(C(=C(C(=C3F)F)F)F)F
C18BF15
MFCD00269813
tris pentafluorophenyl borane, tris perfluorophenyl borane, tris pentafluorophenyl boron, tris 2,3,4,5,6-pentafluorophenyl borane, unii-i3wu5e2578, tris pentafluorophenyl-borane, borane, tris pentafluorophenyl, borane, tris 2,3,4,5,6-pentafluorophenyl, perfluorotriphenylboron, zlchem 1272
tris(2,3,4,5,6-pentafluorophenyl)borane
Safety and Handling

Safety and Handling

TSCA : Yes

Documents