Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(pentafluorophenyl)borane, 97%

Chemical Identifiers

Chemical Name or Material Tris(pentafluorophenyl)borane
CAS 1109-15-5
Molecular Formula C18BF15
Formula Weight 511.99
MDL Number MFCD00269813
UN Number UN2811
Beilstein 2931347
Merck Index 14,9755
Synonym Perfluorotriphenylborane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1805403

 
alfa aesar™
L1805403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1805406

 
alfa aesar™
L1805406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 511.99
Chemical Name or Material Tris(pentafluorophenyl)borane
Assay 97%
Molecular Formula C18BF15
MDL Number MFCD00269813
CAS 1109-15-5
UN Number UN2811
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage −30° to −10°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA TSCA
Beilstein 2931347
Merck Index 14,9755
Synonym Perfluorotriphenylborane
Melting Point 126° to 130°C
Quantity 1g

Applications
Tris(pentafluorophenyl)borane is used as an excellent activator component in homogeneous Ziegler-Natta chemistry. It is also used as a catalyst for reductions, alkylations, hydrometallation reactions and catalyzed aldol-type reactions. It is a useful Lewis acid and catalyzes hydrosilylation of aldehydes. Further, it is used in olefin polymerization catalysis. It serves as a reagent in the preparation of organometallic complexes. In addition to this, it is useful as polymerization catalysts and used to study the heterolytic cleavage of dihydrogen in associated with tri-tert-butylphosphine.

Solubility
Soluble in hexane, chloroform, dichloromethane, toluene, and polar solvents.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and trimethylaluminum.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

−30° to −10°C