Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris[N,N-bis(trimethylsilyl)amide]yttrium(III), 98+%

Chemical Identifiers

Chemical Name or Material Tris[N,N-bis(trimethylsilyl)amide]yttrium(III)
CAS 41836-28-6
Linear Formula [[(CH3)3Si]2N]3Y
Formula Weight 570.06
MDL Number MFCD00210649
UN Number UN3396
Synonym Yttrium(III) tris[N,N-bis(trimethylsilyl)amide]
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3957803

  SDS
alfa aesar™
3957803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3957806

  SDS
alfa aesar™
3957806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 570.06
Boiling Point 105°C (at 10-4mmHg) (subl.)
Packaging Ampule
Quantity 1g
MDL Number MFCD00210649
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P309-P310-P402+P404
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.
Sensitivity Moisture Sensitive
Physical Form Powder
Melting Point 180° to 184°C
Chemical Name or Material Tris[N,N-bis(trimethylsilyl)amide]yttrium(III)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H261-H228-H314-H318
In contact with water releases flammable gas.
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.
DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, WATER-REACTIVE, FLAMMABLE
Assay ≥98%
UN Number UN3396
Linear Formula [[(CH3)3Si]2N]3Y
Color White
CAS 41836-28-6
Synonym Yttrium(III) tris[N,N-bis(trimethylsilyl)amide]
Storage Note 1
  • Ampouled under argon

  • Ampouled under argon

Safety and Handling

Danger

GHS P Statement
P210-P280-P305+P351+P338-P309-P310-P402+P404
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.

GHS H Statement
H261-H228-H314-H318
In contact with water releases flammable gas.
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.

  • Ampouled under argon