Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris[N,N-bis(trimethylsilyl)amide]erbium(III), 98%

Chemical Identifiers

Chemical Name or Material Tris[N,N-bis (trimethylsilyl)amide]erbium (III)
CAS 103457-72-3
Molecular Formula C18H54ErN3Si6
Linear Formula [[(CH3)3Si]2N]3Er
Formula Weight 648.41
MDL Number MFCD00271002
UN Number UN3396
Synonym Erbium(III) tris[N,N-bis(trimethylsilyl)amide]
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4442103

  SDS
alfa aesar™
4442103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4442106

  SDS
alfa aesar™
4442106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 648.41
Quantity 1g
Linear Formula [[(CH3)3Si]2N]3Er
Color Pink
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P309-P310-P402+P404
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.
Health Hazard 2 GHS H Statement
H261-H228-H314-H318
In contact with water releases flammable gas.
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Melting Point 166°C
Health Hazard 1 Danger
Chemical Name or Material Tris[N,N-bis (trimethylsilyl)amide]erbium (III)
Physical Form Powder
UN Number UN3396
DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, WATER-REACTIVE, FLAMMABLE
Molecular Formula C18H54ErN3Si6
Synonym Erbium(III) tris[N,N-bis(trimethylsilyl)amide]
Assay 98%
MDL Number MFCD00271002
Storage Note 1
  • Packaged under Argon
Sensitivity Moisture Sensitive
CAS 103457-72-3

  • Packaged under Argon

Safety and Handling

Danger

GHS P Statement
P210-P280-P305+P351+P338-P309-P310-P402+P404
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Store in a closed container.

GHS H Statement
H261-H228-H314-H318
In contact with water releases flammable gas.
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.

  • Packaged under Argon