missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris[N,N-bis(trimethylsilyl)amide]erbium(III), 98%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 4442106

 View more versions of this product

Catalog No. AA4442106



Description

Description

  • Packaged under Argon
Specifications

Specifications

Tris[N,N-bis (trimethylsilyl)amide]erbium (III)
Pink
5g
C18H54ErN3Si6
MFCD00271002
acmc-20al2x
C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.[Er+3]
648.42
648.41
98%
Moisture sensitive
166°C
103457-72-3
[[(CH3)3Si]2N]3Er
UN3396
ZVNGWTZZRANJAO-UHFFFAOYSA-N
bis(trimethylsilyl)azanide;erbium(3+)
6098739
Powder- Packaged under argon

Chemical Identifiers

103457-72-3
648.42
ZVNGWTZZRANJAO-UHFFFAOYSA-N
6098739
C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.[Er+3]
C18H54ErN3Si6
MFCD00271002
acmc-20al2x
bis(trimethylsilyl)azanide;erbium(3+)
Safety and Handling

Safety and Handling

GHS H Statement
H261-H228-H314-H318
In contact with water releases flammable gas.
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P223-P231+P232-P240-P241-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H228-H261-H314

DOTInformation : Hazard Class: 4.3; Packaging Group: I

TSCA : No

storageNote1 :

  • Packaged under Argon

Recommended Storage : Ambient temperatures

Documents