missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris[N,N-bis(trimethylsilyl)amide]cerium(III), 98%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 4436214

 View more versions of this product

Catalog No. AA4436214



Specifications

Specifications

Tris[N,N-bis (trimethylsilyl)amide]cerium (III)
Yellow
41836-21-9
[[(CH3)3Si]2N]3Ce
UN3396
Soluble in water,protic solvents.
C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.[Ce+3]
621.277
621.27
98%
Moisture sensitive
25g
C18H54CeN3Si6
MFCD03411241
bis trimethylsilyl azanide; cerium 3+
DPCZDYUESIKMSU-UHFFFAOYSA-N
bis(trimethylsilyl)azanide;cerium(3+)
6093781
Powder

Chemical Identifiers

41836-21-9
621.277
DPCZDYUESIKMSU-UHFFFAOYSA-N
6093781
C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.[Ce+3]
C18H54CeN3Si6
MFCD03411241
bis trimethylsilyl azanide; cerium 3+
bis(trimethylsilyl)azanide;cerium(3+)
Safety and Handling

Safety and Handling

GHS H Statement
H261-H228-H314-H318
In contact with water releases flammable gas.
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P223-P231+P232-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c

H228-H261-H314-H335

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, WATER-REACTIVE, FLAMMABLE

TSCA : No

Recommended Storage : Ambient temperatures

Documents