Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(n-butylcyclopentadienyl)yttrium(III), 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Tris(n-butylcyclopentadienyl)yttrium(III)
CAS 312739-77-8
Formula Weight 452.52
MDL Number MFCD00145473
UN Number UN1993
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3957703

  SDS
alfa aesar™
3957703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3957706

  SDS
alfa aesar™
3957706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 452.52
Physical Form Liquid, Ampouled Under Argon
Health Hazard 1 Warning
UN Number UN1993
Chemical Name or Material Tris(n-butylcyclopentadienyl)yttrium(III)
CAS 312739-77-8
Assay Percent Notes (REO)
MDL Number MFCD00145473
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
Quantity 1g
Assay 99.9%
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Sensitivity Air & Moisture Sensitive
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapor.