Please login to your online account to display your discounted pricing

Tris Molecular Biology MP Biomedicals

$362.41 - $362.41

Chemical Identifiers

Chemical Name or Material Tris
CAS 77-86-1
Purity Grade Molecular Biology Grade
Molecular Formula C4H11NO3
Formula Weight 121.14
Synonym Trometamol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19485591

 
mp biomedicals
19485591
1kg Each for $362.41
Description & Specifications

Specifications

Quantity 1kg
Purity Grade Molecular Biology Grade
Formula Weight 121.14
Chemical Name or Material Tris
Synonym Trometamol
CAS 77-86-1
Molecular Formula C4H11NO3