Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(isopropylcyclopentadienyl)terbium(III), 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Tris(isopropylcyclopentadienyl)terbium(III)
CAS 312696-25-6
Formula Weight 480.45
MDL Number MFCD00798537
UN Number UN3181
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4050603

  SDS
alfa aesar™
4050603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 480.45
Packaging Ampule
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN3181
Assay 99.9%
Sensitivity Air & Moisture Sensitive
Assay Percent Notes (REO)
MDL Number MFCD00798537
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Physical Form Crystalline
CAS 312696-25-6
Storage Note 1
  • Ampouled under argon
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
Chemical Name or Material Tris(isopropylcyclopentadienyl)terbium(III)

  • Ampouled under argon

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

  • Ampouled under argon