Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(isopropylcyclopentadienyl)lanthanum(III), 99.9% (REO)

$113.62 - $113.62

Chemical Identifiers

Chemical Name or Material Tris(isopropylcyclopentadienyl)lanthanum(III)
CAS 68959-87-5
Formula Weight 460.43
MDL Number MFCD00145482
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3932303

  SDS
alfa aesar™
3932303
1g Each for $113.62
Description & Specifications

Specifications

Physical Form Liquid, Ampouled Under Argon
Formula Weight 460.43
MDL Number MFCD00145482
Sensitivity Air & Moisture Sensitive
Health Hazard 1 Warning
CAS 68959-87-5
Chemical Name or Material Tris(isopropylcyclopentadienyl)lanthanum(III)
Color Colorless to pale yellow
Assay 99.9%
Quantity 1g
Assay Percent Notes (REO)
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.