Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(hydroxymethyl)aminomethane hydrochloride, Molecular Biology Grade, 99+%

$58.91 - $58.91

Chemical Identifiers

Chemical Name or Material Tris(hydroxymethyl)aminomethane hydrochloride
CAS 1185-53-1
Purity Grade Molecular Biology Grade
Molecular Formula C4H11NO3·HCl
Formula Weight 157.6
MDL Number MFCD00012590
Beilstein 3675235
Synonym TRIS hydrochloride; 2-Amino-2-hydroxymethyl-1,3-propanediol hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ6723322

  SDS
alfa aesar™
J6723322
100g Each for $58.91
Description & Specifications

Specifications

Formula Weight 157.6
Chemical Name or Material Tris(hydroxymethyl)aminomethane hydrochloride
Melting Point ≈152°C
pH 3.5 to 5.0
Sensitivity Hygroscopic
Purity Grade Molecular Biology Grade
Physical Form Powder
Assay ≥99%
Molecular Formula C4H11NO3·HCl
MDL Number MFCD00012590
CAS 1185-53-1
Beilstein 3675235
Synonym TRIS hydrochloride; 2-Amino-2-hydroxymethyl-1,3-propanediol hydrochloride
Color White
Quantity 100g
TSCA TSCA

  • For adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols
  • Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications