missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris(hydroxymethyl)aminomethane Hydrochloride (Crystalline/Certified) Reagent Grade, Fisher Chemical

Manufacturer:  Fisher Chemical FLO4997100

Catalog No. O4997-100




Description

Description

Specifications

Specifications

Tris(hydroxymethyl)aminomethane Hydrochloride
≥95 %
NH2C(CH2OH)3·HCl
2-Amino-2-(hydroxymethyl)-1, 3-propanediol Hydrochloride, Tris Hydrochloride
C(C(CO)(CO)N)O.Cl
157.594
157.6
Solid
Reagent
150°C
White
1185-53-1
C4H12ClNO3
MFCD00012590
QKNYBSVHEMOAJP-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
93573
Pass Test
≥95%
Glass Bottle
3.5 to 5.0
100g

Chemical Identifiers

1185-53-1
157.594
QKNYBSVHEMOAJP-UHFFFAOYSA-N
93573
C(C(CO)(CO)N)O.Cl
C4H12ClNO3
MFCD00012590
2-Amino-2-(hydroxymethyl)-1, 3-propanediol Hydrochloride, Tris Hydrochloride
2-amino-2-(hydroxymethyl)propane-1,3-diol;hydrochloride
Safety and Handling

Safety and Handling

SDS Documents