Please login to your online account to display your discounted pricing

Tris(hydroxymethyl)aminomethane (Crystalline/Laboratory), Fisher Chemical


Quantity 3kg
Packaging Poly Pail

Manufacturer: fisher scientific company  T3703

 View more versions of this product

Catalog No. T370-3

  • / Each
 

Description & Specifications

Chemical Identifiers

Boiling Point 219°C
Melting Point 168.5°C
Purity Grade Laboratory Grade
Physical Form Powder/Solid
Formula Weight 121.14
Quantity 3kg
Color White
Molecular Formula C4H11NO3
pH 10.4
Synonym 2-Amino-2-(hydroxymethyl)-1, 3-Propanediol Trimethylol Aminomethane
MDL Number MFCD00004679
ChemAlert Storage Symbol Gray
Identification Pass Test
Packaging Poly Pail
CAS 77-86-1
Chemical Name or Material Tris(hydroxymethyl)aminomethane
Linear Formula H2NC(CH2OH)3

Specifications

Chemical Name or Material Tris(hydroxymethyl)aminomethane
CAS 77-86-1
Purity Grade Laboratory Grade
Physical Form Powder/Solid
Molecular Formula C4H11NO3
Linear Formula H2NC(CH2OH)3
Formula Weight 121.14
MDL Number MFCD00004679
Synonym 2-Amino-2-(hydroxymethyl)-1, 3-Propanediol Trimethylol Aminomethane
Accessories
 

SDS & Safety Documents