missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

Tris(hydroxymethyl)aminomethane (Crystalline/Laboratory), Fisher Chemical™
Manufacturer: Fisher Chemical T370500
Specifications
Tris(hydroxymethyl)aminomethane | |
C4H11NO3 | |
MFCD00004679 | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
2-amino-2-(hydroxymethyl)propane-1,3-diol | |
6503 | |
121.14 | |
Powder/Solid | |
Glass Bottle | |
168.5°C | |
White |
77-86-1 | |
H2NC(CH2OH)3 | |
2-Amino-2-(hydroxymethyl)-1, 3-Propanediol Trimethylol Aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trizma™, Trometamol | |
C(C(CO)(CO)N)O | |
121.136 | |
CHEBI:9754 | |
Pass Test | |
Laboratory | |
219°C | |
10.4 | |
500g |
Chemical Identifiers
77-86-1 | |
121.136 | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
6503 | |
2-amino-2-(hydroxymethyl)propane-1,3-diol |
C4H11NO3 | |
MFCD00004679 | |
2-Amino-2-(hydroxymethyl)-1, 3-Propanediol Trimethylol Aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trizma™, Trometamol | |
CHEBI:9754 | |
C(C(CO)(CO)N)O |
Safety and Handling
