Please login to your online account to display your discounted pricing

Tris(hydroxymethyl)aminomethane (Crystalline/Laboratory), Fisher Chemical

$462.46 - $2,124.31

Chemical Identifiers

Chemical Name or Material Tris(hydroxymethyl)aminomethane
CAS 77-86-1
Purity Grade Laboratory Grade
Physical Form Powder/Solid
Molecular Formula C4H11NO3
Linear Formula H2NC(CH2OH)3
Formula Weight 121.14
MDL Number MFCD00004679
Synonym 2-Amino-2-(hydroxymethyl)-1, 3-Propanediol Trimethylol Aminomethane
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

T370-500

  SDS
fisher scientific company
T370500
Glass Bottle 500g Each for $462.46
Case of 6 Each for $2,774.76

T370-3

  SDS
fisher scientific company
T3703
Poly Pail 3kg Each for $2,124.31
Description & Specifications

Specifications

Purity Grade Laboratory Grade
Formula Weight 121.14
Physical Form Powder/Solid
Melting Point 168.5°C
Boiling Point 219°C
Quantity 500g
Color White
pH 10.4
Molecular Formula C4H11NO3
Packaging Glass Bottle
ChemAlert Storage Symbol Gray
MDL Number MFCD00004679
Identification Pass Test
Synonym 2-Amino-2-(hydroxymethyl)-1, 3-Propanediol Trimethylol Aminomethane
Chemical Name or Material Tris(hydroxymethyl)aminomethane
Linear Formula H2NC(CH2OH)3
CAS 77-86-1
Safety and Handling