missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(hydroxymethyl)aminomethane, ACS, 99.8-100.1% (Assay, dried basis)

Manufacturer:  Alfa Aesar™ 31801A1

 View more versions of this product

Catalog No. AA31801A1



Specifications

Specifications

Tris(hydroxymethyl)aminomethane
ACS Grade
(Assay, dried basis)
MFCD00004679
14,9772
LENZDBCJOHFCAS-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol
6503
121.14
1.353g/mL
219°C to 220°C (10mmHg)
Hygroscopic
77-86-1
99.8 to 100.1%
C4H11NO3
741883
2-Amino-2-hydroxymethyl-1,3-propanediol; TRIS
C(C(CO)(CO)N)O
121.136
CHEBI:9754
Crystalline
171°C to 172°C
1kg

Chemical Identifiers

77-86-1
121.136
LENZDBCJOHFCAS-UHFFFAOYSA-N
6503
2-amino-2-(hydroxymethyl)propane-1,3-diol
C4H11NO3
MFCD00004679
2-Amino-2-hydroxymethyl-1,3-propanediol; TRIS
CHEBI:9754
C(C(CO)(CO)N)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : TY2900000

TSCA : TSCA

SDS Documents