Please login to your online account to display your discounted pricing

TRIS Hydrochloride, Ultrapure Bioreagent, J.T.Baker™


High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

Quantity 250g
Packaging Poly Bottle

Manufacturer: avantor performance  410304

 View more versions of this product

Catalog No. 02004518

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 157.6
Chemical Name or Material TRIS Hydrochloride
Molecular Formula NH2C(CH2OH)3·HCl
Packaging Poly Bottle
CAS 1185-53-1
Purity Grade Ultrapure Bioreagent
Quantity 250g

Specifications

Chemical Name or Material TRIS Hydrochloride
CAS 1185-53-1
Purity Grade Ultrapure Bioreagent
Molecular Formula NH2C(CH2OH)3·HCl
Formula Weight 157.6

    For Liquid Chromatography and Molecular Biology Applications