missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris Hydrochloride (Small White Flakes/Molecular Biology), Fisher BioReagents

For electrophoresis, molecular biology, and cell culture applications

Manufacturer:  Fisher Scientific Company BP153500

 View more versions of this product

Catalog No. BP153-500



 

Specifications Description & Specifications

Specifications

0.05 max. (1M solution) at 280nm
1185-53-1
1ppm max.
Molecular Biology Grade
Solid
≥99 %
1ppm max.
157.6
Pass Test
Pass Test
2ppm max.
1ppm max.
Protease free
3.5 to 5.0
500g
Tris Hydrochloride
10ppm max.
≤0.001%
DNase-, RNase- and Protease-Free
≥99%
C4H11NO3 · HCl
H2NC(CH2OH)3·HCl
DNase free
Tris(hydroxymethyl)aminomethane Hydrochloride
5ppm max.
≤10ppm
Poly Bottle
150°C
White

Chemical Identifiers

1185-53-1
≥99 %
H2NC(CH2OH)3·HCl
Tris(hydroxymethyl)aminomethane Hydrochloride
≥99%
Solid
C4H11NO3 · HCl
157.6
Tris Hydrochloride
Molecular Biology Grade, DNase-, RNase- and Protease-Free

Safety and Handling

SDS & Safety Documents

Recommended Storage : Keep in a dry, cool and well-ventilated place. Keep container tightly closed.