Please login to your online account to display your discounted pricing

TRIS Hydrochloride, Made from Tromethamine USP, Biotech Reagent Grade, J.T.Baker™


Produced under GMP guidelines, but for which there is no USP/NF compendia for the product

Quantity 500g
Packaging Poly bottle

Manufacturer: avantor performance  410601

 View more versions of this product

Catalog No. 02004520

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 157.6
Melting Point 147° to 153°C
Purity Grade Notes GMP manufactured product
Chemical Name or Material TRIS Hydrochloride
CAS 1185-53-1
Molecular Formula NH2C(CH2OH)3·HCl
Name Note Made from Tromethamine USP
Specific Gravity 1L = 0.00kg
Purity Grade Biotech Reagent Grade
Quantity 500g
Packaging Poly bottle

Specifications

Chemical Name or Material TRIS Hydrochloride
CAS 1185-53-1
Name Note Made from Tromethamine USP
Purity Grade Biotech Reagent Grade
Purity Grade Notes GMP manufactured product
Molecular Formula NH2C(CH2OH)3·HCl
Formula Weight 157.6