Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris-Glycine-SDS Large Precast Gel, 2D, 8-16%, 20x20cm, 22.3x20cm (back), low fluor., 1 well, for Bio-Rad™ systems


Manufacturer: alfa aesar™  J67598KZ

 View more versions of this product

Catalog No. AAJ67598KZ

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Storage Requirements 2° to 8°C
Quantity 6 Each
Boiling Point 100°C
Product Type Tris-Glycine-SDS Large Precast Gel
Color Colorless
Odor Odorless
Format Solid