Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris-Glycine-SDS EL Precast Gel, 2D, 4-20%, 21.2x27.6cm, 21.7x27.6cm (back), low fluor., bind silane, Ettan Dalt™ systems


Manufacturer: alfa aesar™  J67689SA

 View more versions of this product

Catalog No. AAJ67689SA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Boiling Point 100°C
Quantity 2 Each
Storage Requirements 2° to 8°C
Product Type Tris-Glycine-SDS EL Precast Gel
Color Colorless
Odor Odorless
Format Solid