Alfa Aesar™ Tris-Glycine Large Precast Gel, 2D, 10-20%, 20x20cm, 22.3x20cm (back), 1mm thick, 1 well, for Bio-Rad™ systems
Specifications
Product Type Tris-Glycine Large Precast Gel
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ67526KZ Alfa Aesar™
J67526KZ
6 Each N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAJ67526SA Alfa Aesar™
J67526SA
2 Each N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications
Specifications
Tris-Glycine Large Precast Gel