Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris-Glycine Large Precast Gel, 2D, 10-20%, 20x20cm, 22.3x20cm (back), 1mm thick, 1 well, for Bio-Rad™ systems


Manufacturer: alfa aesar™  J67526SA

 View more versions of this product

Catalog No. AAJ67526SA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 2 Each
Storage Requirements 2° to 8°C
Boiling Point 100°C
Product Type Tris-Glycine Large Precast Gel
Color Colorless
Format Solid
Odor Odorless