Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(ethylthio)methane, 98%

$203.91 - $203.91

Chemical Identifiers

Chemical Name or Material Tris(ethylthio)methane
CAS 6267-24-9
Molecular Formula C7H16S3
Linear Formula HC(SCH2CH3)3
Formula Weight 196.38g/mol
MDL Number MFCD00015179
Beilstein 1737016
Synonym Ethyl orthothioformate; Triethyl trithioorthoformate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0290714

 
alfa aesar™
L02907-14
25g Each for $203.91
Description & Specifications

Specifications

Quantity 25g
Boiling Point 91° to 92°C (0.5mmHg)
Flash Point >63°C (145°F)
Odor Unpleasant
Quantity 25g
Chemical Name or Material Tris(ethylthio)methane
CAS 6267-24-9
Assay 98%
Molecular Formula C7H16S3
Linear Formula HC(SCH2CH3)3
Formula Weight 196.38g/mol
MDL Number MFCD00015179
Beilstein 1737016
Synonym Ethyl orthothioformate; Triethyl trithioorthoformate
Health Hazard 1 Warning
TSCA TSCA