Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(ethylenediamine)rhodium(III) chloride trihydrate, Rh 23.2%

Chemical Identifiers

Chemical Name or Material Tris(ethylenediamine)rhodium(III) chloride trihydrate
CAS 15004-86-1
Linear Formula Rh(H2NCH2CH2NH2)3Cl3·3H2O
Formula Weight 443.61 (389.56 Anhydrous)
MDL Number MFCD00149661
Synonym Trichlorotris(ethylenediamine)rhodium(III)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1055302

  SDS
alfa aesar™
1055302
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1055303

  SDS
alfa aesar™
1055303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1055306

  SDS
alfa aesar™
1055306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline/Fused Lump, Crystals Fuse Over Time
Formula Weight 443.61 (389.56 Anhydrous)
Chemical Name or Material Tris(ethylenediamine)rhodium(III) chloride trihydrate
Linear Formula Rh(H2NCH2CH2NH2)3Cl3·3H2O
MDL Number MFCD00149661
CAS 15004-86-1
Synonym Trichlorotris(ethylenediamine)rhodium(III)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Color White
Melting Point 280°C (dec.)
Concentration Rh 23.2%
Quantity 0.5g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.