missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(ethylenediamine)rhodium(III) chloride trihydrate, Rh 23.2%

Manufacturer:  Alfa Aesar™ 1055303

 View more versions of this product

Catalog No. AA1055303



Specifications

Specifications

Tris(ethylenediamine)rhodium(III) chloride trihydrate
280°C (decomposition)
15004-86-1
Rh(H2NCH2CH2NH2)3Cl3·3H2O
Trichlorotris(ethylenediamine)rhodium(III)
HHSLUQMEKOPKJG-UHFFFAOYSA-K
443.61 (389.56 Anhydrous)
White
1g
C6H30Cl3N6O3Rh
MFCD00149661
Rh 23.2%
443.60
Crystalline/Fused Lump, Crystals Fuse Over Time

Chemical Identifiers

15004-86-1
443.60
HHSLUQMEKOPKJG-UHFFFAOYSA-K
C6H30Cl3N6O3Rh
MFCD00149661
Trichlorotris(ethylenediamine)rhodium(III)
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents