Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(ethylenediamine)iron(III) sulfate

Chemical Identifiers

Chemical Name or Material Tris(ethylenediamine)iron(III) sulfate
Formula Weight 760.38
MDL Number MFCD00035682
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3112309

 
alfa aesar™
3112309
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3112318

 
alfa aesar™
3112318
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 760.38
Chemical Name or Material Tris(ethylenediamine)iron(III) sulfate
Physical Form Powder
MDL Number MFCD00035682
Quantity 10g