Alfa Aesar™ Tris(ethylenediamine)chromium(III) chloride hemiheptahydrate
Manufacturer:  Alfa Aesar™ 2412703
 View more versions of this product
Catalog No. AA2412703
Specifications
Specifications
Tris(ethylenediamine)chromium(III) chloride hemiheptahydrate
1g
Cr(H2NCH2CH2NH2)3Cl3·3.5H2O
Trichlorotris(ethylenediamine)chromium(III)
Crystalline
Yellow to Orange
16165-32-5
MFCD00150160
401.71 (338.65 Anhydrous)
Chemical Identifiers
16165-32-5
MFCD00150160
Trichlorotris(ethylenediamine)chromium(III)
Safety and Handling
Safety and Handling
GHS H Statement
H317
May cause an allergic skin reaction.
GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
SDS Documents