Please login to your online account to display your discounted pricing

Alfa Aesar™ Tris(ethylenediamine)chromium(III) chloride hemiheptahydrate

$73.74 - $741.93

Chemical Identifiers

Chemical Name or Material Tris(ethylenediamine)chromium(III) chloride hemiheptahydrate
CAS 16165-32-5
Linear Formula Cr(H2NCH2CH2NH2)3Cl3·3.5H2O
Formula Weight 401.71 (338.65 Anhydrous)
MDL Number MFCD00150160
Synonym Trichlorotris(ethylenediamine)chromium(III)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA2412706

 
alfa aesar™
2412706
5g Each for $73.74

AA2412703

 
alfa aesar™
2412703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA2412714

 
alfa aesar™
2412714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA2412722

 
alfa aesar™
2412722
100g Each for $741.93
Description & Specifications

Specifications

Formula Weight 401.71 (338.65 Anhydrous)
Health Hazard 3 GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Tris(ethylenediamine)chromium(III) chloride hemiheptahydrate
Physical Form Crystalline
Linear Formula Cr(H2NCH2CH2NH2)3Cl3·3.5H2O
MDL Number MFCD00150160
CAS 16165-32-5
Synonym Trichlorotris(ethylenediamine)chromium(III)
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H317
May cause an allergic skin reaction.
Color Yellow to orange
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H317
May cause an allergic skin reaction.