Please login to your online account to display your discounted pricing

TE Buffer, Tris-EDTA, 10X Solution, pH 7.6, Molecular Biology, Fisher BioReagents


Quantity 500mL
Packaging Poly Bottle

Manufacturer: fisher scientific company  BP2475500

 View more versions of this product

Catalog No. BP2475500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Tris-EDTA (TE), 10X Solution
Name Note pH 7.6
CAS 77-86-1
ChemAlert Storage Symbol Gray
Health Hazard 2 CAUTION!
CAS 6381-92-6
Health Hazard 3 Emergency Overview
May cause eye, skin, and respiratory tract irritation. Avoid contact with skin and eyes. Do not breathe dust. Do not breathe vapors or spray mist. Wash off immediately with soap and plenty of water removing all contaminated clothes and shoes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Remove from exposure, lie down. Move to fresh air. If breathing is difficult, give oxygen. If not breathing, give artificial respiration. If not breathing, give artificial respiration. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:0
Recommended Storage RT
CAS 7647-01-0
CAS 7732-18-5
Purity Grade Molecular Biology Grade
Physical Form Liquid
DNase DNase free
Synonym TE
Packaging Poly Bottle
Protease Protease free
Filtered Through Filtered through a 5-micron filter and autoclaved.
pH 7.4 to 8.1
Color Colorless
Quantity 500mL

Specifications

Chemical Name or Material Tris-EDTA (TE), 10X Solution
Name Note pH 7.6
CAS 77-86-1
CAS 6381-92-6
CAS 7647-01-0
CAS 7732-18-5
Purity Grade Molecular Biology Grade
Synonym TE

  • Save time and simplify your buffer preparation step by using Fisher BioReagents 10X TE buffer solution - simply dilute as needed
  • 10X solution contains 100mM tris and 10mM EDTA
  • To prepare a 1X solution, mix one volume of Fisher BioReagents 10X TE solution with 9 volumes of ultrapure water, such as BP2819
  • Filtered through a 5 micron filter and autoclaved
  • Tested for the absence of DNase, RNase, and protease
  • TE buffer is routinely used for resuspending nucleic acid samples
  • Intended for laboratory research use only

Accessories
 

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Avoid contact with skin and eyes. Do not breathe dust. Do not breathe vapors or spray mist. Wash off immediately with soap and plenty of water removing all contaminated clothes and shoes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Remove from exposure, lie down. Move to fresh air. If breathing is difficult, give oxygen. If not breathing, give artificial respiration. If not breathing, give artificial respiration. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:0

CAUTION!

RT

SDS & Safety Documents