Please login to your online account to display your discounted pricing

TE Buffer, Tris-EDTA, 10X Solution, pH 7.6, Molecular Biology, Fisher BioReagents

$49.85 - $125.32

Chemical Identifiers

Chemical Name or Material Tris-EDTA (TE), 10X Solution
Name Note pH 7.6
CAS 77-86-1
CAS 6381-92-6
CAS 7647-01-0
CAS 7732-18-5
Purity Grade Molecular Biology Grade
Synonym TE
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BP2475500

  SDS
fisher scientific company
BP2475500
Poly Bottle 500mL Each for $64.59
Case of 6 Each for $387.54

BP2475100

  SDS
fisher scientific company
BP2475100
Poly Bottle 100mL Each for $49.85
Case of 6 Each for $299.10

BP24751

  SDS
fisher scientific company
BP24751
Nalgene Poly Bottle 1L Each for $125.32
Case of 6 Each for $751.92
Description & Specifications

Specifications

Purity Grade Molecular Biology Grade
Quantity 500mL
Filtered Through Filtered through a 5-micron filter and autoclaved.
pH 7.4 to 8.1
Color Colorless
Physical Form Liquid
Conductivity [NULL]
DNase DNase free
Protease Protease free
CAS 7732-18-5
Name Note pH 7.6
CAS 77-86-1
Synonym TE
Health Hazard 2 CAUTION!
CAS 6381-92-6
CAS 7647-01-0
Chemical Name or Material Tris-EDTA (TE), 10X Solution
ChemAlert Storage Symbol Gray
Health Hazard 3 Emergency Overview
May cause eye, skin, and respiratory tract irritation. Avoid contact with skin and eyes. Do not breathe dust. Do not breathe vapors or spray mist. Wash off immediately with soap and plenty of water removing all contaminated clothes and shoes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Remove from exposure, lie down. Move to fresh air. If breathing is difficult, give oxygen. If not breathing, give artificial respiration. If not breathing, give artificial respiration. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:0
Packaging Poly Bottle
Recommended Storage RT

Tris–EDTA (TE) is used for suspending nucleic acid samples.

Safety and Handling
 

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Avoid contact with skin and eyes. Do not breathe dust. Do not breathe vapors or spray mist. Wash off immediately with soap and plenty of water removing all contaminated clothes and shoes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Remove from exposure, lie down. Move to fresh air. If breathing is difficult, give oxygen. If not breathing, give artificial respiration. If not breathing, give artificial respiration. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:0

CAUTION!

RT