missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tris EDTA buffer, 1X, Molecular Biology Grade DFS Item

10mM Tris-HCl containing 1mM EDTA-Na2.

Manufacturer:  Promega Corportaion V6232

 View more versions of this product

Catalog No. PR-V6232



Specifications

Specifications

500mL

Chemical Identifiers

0.0
icon-pdf.png
Safety and Handling

Safety and Handling

Recommended Storage : 15 to 30°C

Documents
Promotions

Promotions